Produktname
Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride protease substrate
Produktnummer
b7260
Produktmarke
sigma
CAS-Nummer
83701-04-6
Molekulargewicht
471.94
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White |
| APPEARANCE (FORM) | Powder |
| PURITY (TLC AREA %) | ≥ 99 % |
| SPECIFIC ROTATION | 51.0 - 55.0 Degrees |
| CONCENTRATION | C=1 IN DIMETHYL SULFOXIDE, 25 C |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 20MG/ML IN DIMETHYL SULFOXIDE:METHANOL (1:1) |
| CARBON CONTENT | 57.3 - 59.4 % |
| NITROGEN CONTENT | 14.5 - 15.2 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |