Produktname
L-Cystein -hydrochlorid anhydrous, ≥98% (TLC)
Produktnummer
c1276
Produktmarke
sigma
CAS-Nummer
52-89-1
Molekulargewicht
157.62
Test | Spezifikation |
|---|---|
| Appearance (Color) | White |
| Appearance (Form) | Powder |
| Solubility (Color) | Colorless |
| Solubility (Turbidity) | Clear |
| at 50 mg/ml in H2O | |
| Infrared spectrum | Conforms to Structure |
| Carbon | 22.4 - 23.3 % |
| Nitrogen | 8.6 - 9.2 % |
| Purity (TLC) | ≥98 % |
| Recommended Retest Period | ------------------------------ |
| 4 years |