Produktname
Choleratoxin aus Vibrio cholerae lyophilized powder
Produktnummer
c3012
Produktmarke
sigma
CAS-Nummer
9012-63-9
Molekulargewicht
Test | Spezifikation |
|---|---|
| SDS ELECTROPHORESIS | CONSISTENT WITH LITERATURE (METHODS ENZYMOLOGY, VOL. 165, P. 169 (1988)) |
| VIAL CONTENT | REPORT RESULT |
| NOTE | LYOPHILIZED POWDER CONTAINING TRIS BUFFER SALTS, SODIUM CHLORIDE, SODIUM AZIDE AND EDTA-DISODIUM SALT |
| SHELF LIFE | 3 YEARS |
| DOCUMENT # C3012/04/19/00/3 |