Produktname
4-Chlor-3,5-Dimethylphenol ≥99%
Produktnummer
c4394
Produktmarke
sigma
CAS-Nummer
88-04-0
Molekulargewicht
156.61
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO OFF-WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS TO VERY FAINT YELLOW SOLUTION AT 100MG/ML IN ETHANOL |
| IR SPECTRUM | CONSISTENT WITH STRUCTURE |
| PURITY BY GAS CHROMATOGRAPHY | NLT 98% |
| DOCUMENT # C4394/02/19/03/2 |