Produktname
L-Carnitin Inneres Salz from equine muscle, ~98%
Produktnummer
c7393
Produktmarke
sigma
CAS-Nummer
541-15-1
Molekulargewicht
161.20
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS SOLUTION AT 50 MG/ML OF WATER |
| IR SPECTRUM | CONSISTENT WITH STRUCTURE |
| SPECIFIC ROTATION | CONSISTENT WITH OPTICAL ISOMER |
| WATER BY KARL FISCHER | < OR = 3% |
| ELEMENTAL ANALYSIS | 8.2 TO 9.5% NITROGEN |
| PURITY BY THIN LAYER CHROMATOGRAPHY | APPROX 98% |
| PROTON NMR SPECTRUM | CONSISTENT WITH STRUCTURE |
| DOCUMENT # | C7393/06/25/07/1 |