Produktname
S-Carboxymethyl-L-Cystein
Produktnummer
c7757
Produktmarke
sigma
CAS-Nummer
638-23-3
Molekulargewicht
179.19
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS SOLUTION AT 200MG PLUS 4ML OF 1M AMMONIUM HYDROXIDE |
| CARBON | 32.8 TO 34.2% |
| NITROGEN | 7.5 TO 8.1% |
| PURITY BY THIN LAYER CHROMATOGRAPHY | NOT LESS THAN 98% |
| APPROVED HAM 12/15/92 |