Produktname
L-Carnosin ~99%, crystalline
Produktnummer
c9625
Produktmarke
sigma
CAS-Nummer
305-84-0
Molekulargewicht
226.23
Test | Spezifikation |
|---|---|
| Appearance (Color) | White to Off-White |
| Appearance (Form) | Powder |
| Solubility (Color) | Colorless to Light Yellow |
| 20 mg/ml in Water | |
| Solubility (Turbidity) | Clear to Slightly Hazy |
| Carbon | 46.8 - 48.7 % |
| Nitrogen | 24.2 - 25.2 % |
| Infrared spectrum | Conforms to Structure |
| Purity (TLC) | ≥98 % |