Produktname
Diethylentriamin-pentaessigsäure-dianhydrid
Produktnummer
d6148
Produktmarke
sigma
CAS-Nummer
23911-26-4
Molekulargewicht
357.32
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO YELLOW WITH A TAN CAST POWDER |
| SOLUBILITY | CLEAR LIGHT YELLOW SOLUTION AT 50MG/ML IN DIMETHYL SULFOXIDE |
| PROTON NMR SPECTRUM | CONSISTENT WITH STRUCTURE |
| PURITY BY NAOH TITRATION | NLT 98% |
| DOCUMENT # D6148/04/20/90/1 |