Produktname
Earle-Salze suitable for cell culture, liquid
Produktnummer
e2899
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE | CLEAR SOLUTION |
| PH TEST AT RT | 7.0 - 7.4 |
| OSMOLALITY TEST | 265 - 291 MOSM/KG |
| ENDOTOXIN ASSAY | NMT 1.0 EU/ML |
| GLUCOSE | 0.9 - 1.1 G/L |
| STERILE BY USP | STERILE |
| KEY ELEMENTS BY ICP | CONSISTENT WITH FORMULA |
| CELL CULTURE TEST | PASS |
| EXPIRATION DATE | 24 MONTHS |
| E2899:1/99-1 |