Produktname
Ethylendiamintetraessigsäure -Lösung 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Produktnummer
e8008
Produktmarke
sigma
CAS-Nummer
60-00-4
Molekulargewicht
292.24
Test | Spezifikation |
|---|---|
| Appearance (Turbidity) | Clear |
| Appearance (Form) | Solution |
| pH | 7.1 - 7.7 |
| Osmolality | 262 - 288 mOs/kg |
| Sterility | Pass |
| Endotoxin Level | EU/ml |
| Cell Dissociation Assay | Pass |