Produktname
Dulbecco’s Modified Eagle’s Medium - low glucose With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Produktnummer
f0415
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Appearance (Colour) | Red |
| Appearance (Form) | Clear Solution |
| pH | 7.1 - 7.4 |
| pH Temperature | 20 - 22 °C |
| Osmolality | 300 - 330 mOs/kg |
| Endotoxin | <= 1.5 EU/ml |
| Sterility by current EP/USP | Pass |
| Cell Growth | Pass |
| Cell Cloning | Pass |