Produktname
N-Formyl-L-methionine ≥90% (TLC)
Produktnummer
f3377
Produktmarke
sigma
CAS-Nummer
4289-98-9
Molekulargewicht
177.22
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White |
| APPEARANCE (FORM) | Powder |
| PURITY (TLC AREA %) | ≥ 90 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear to Slightly Hazy |
| SOLUBILITY (METHOD) | 50MG/ML IN METHANOL |
| CARBON CONTENT | 39.9 - 41.5 % |
| NITROGEN CONTENT | 7.6 - 8.2 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |