Produktname
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
Produktnummer
f5135
Produktmarke
sigma
CAS-Nummer
78832-65-2
Molekulargewicht
476.52
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Beige |
| APPEARANCE (FORM) | Powder |
| PURITY (HPLC AREA %) | ≥ 95 % |
| SOLUBILITY (COLOR) | Colorless to Faint Yellow |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 2 MG/ML IN METHANOL |
| CARBON CONTENT | 54.9 - 60.0 % |
| NITROGEN CONTENT | 11.1 - 12.2 % |