Produktname
Fetales Kälberserum USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Produktnummer
f6765
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Mycoplasma (Broth Culture) | None detected |
| Osmolality | 260 - 340 mOsm/kg H2O |
| AVA (9CFR113.53) | BAV (Type 3 and 5) - None detected |
| Bovine Parvovirus - None detected | |
| Blue Tongue Virus - None detected | |
| Bovine Viral Diarrhea - Tested | |
| Cytopathic Effect - None detected | |
| Hemadsorption - None detected | |
| Infectious Bovine Rhinotracheitis -,None detected | |
| Parainfuenza 3 - None detected | |
| Rabies Virus - None detected | |
| Reovirus - None detected | |
| Bovine Respiratory Syncytial Virus -,None detected | |
| Vesicular Stomatitis Virus - None,detected | |
| Progesterone (Post) | Report result |
| Total Protein | 2.5 - 4.5 g% |
| T3 (Post) | Report result |
| T4 (Post) | Report result |
| Chemical Analysis (Cholesterol) | Report result (mg/dL) |
| Hormone Level - Estradiol | Report result |
| Chemical Analysis (Iron) | Report result (mcg/dL) |
| Chemical Analysis (Glucose) | Report result (mg/dL) |
| Hormone Level - Insulin Level | Report result |
| Chemical Analysis (Sodium) | Report result (mEq/L) |
| Hormone Level - Progesterone | Report result |
| Hormone Level - Testosterone | Report result |
| Tetracycline | Report Result |
| Chemical Analysis (Triglyceride) | Report result (mg/dL) |
| Serum Antibody Titer - BRSV | Report result |
| Serum Antibody Titer - BVDV | Report result |
| Serum Antibody Titer - IBR | Report result |
| Serum Antibody Titer - PI3 | Report result |
| Cloning Assay | Pass |
| Mycoplasma (DNA Fluorochrome Stain) | None detected |
| pH | 6.7 - 8.0 |
| Bovine IgG | <= 1 mg/mL |
| Endotoxin | <= 10 EU/mL |
| Serum Performance Test | Pass |
| Cell Line | Record cell lines used FIO |
| Hemoglobin | <= 20 mg% |
| Electrophoretic Profile | Albumin (g/dL) - Record |
| Alpha 1 Globulin (g/dL) - Record | |
| Beta Globulin (g/dL) - Record | |
| Gamma Globulin (g/dL) - Record | |
| T3 (Pre) | Report results (Pre charcoal stripped) |
| T4 (Pre) | Report results (Pre charcoal stripped) |
| Country of Origin | Record |
| Progesterone (Pre) | Report result (Pre charcoal stripped) |
| Appearance | Clear straw to amber colored liquid |
| COMMENTS | |
| Intended Use | For R&D use only. Not for drug, household, or other uses. |
| Storage | Store at -20 C |
| Extended Item Desc. | Fetal Bovine Serum Charcoal Stripped, USA Origin |
| Origin | The material used in this product was collected in the United States. |
| Animals used for collection of serum were USDA inspected and acceptable | |
| for slaughter. | |
| C of A comments | Country of final product processing: United States |
| Label Requirements | DOM: N EXP: Y Format MM/YY |
| Expiration Notes for FG | Expiration Date: 48 months from date of manufacture |
| Stop Lot Date: 46 months from date of manufacture | |
| Label Requirements | Expiration date on the Sigma label is in MM/YY format. As an example: |
| if material expires 31AUG12 the expiration date on the label will be | |
| listed as 08/12. | |
| Use Sigma label stock. | |
| Label Comments | 'Product of XX' must appear on the label where XX will be the Raw Serum's |
| Country of Origin. | |
| Misc. | Create both a BPCS generated COA and a SAP generated COA. |
| Specification | F6765.000000004 |
| Approval Date | 12/19/2024 |