Produktname
Prostaglandin F Synthase human recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Produktnummer
f7807
Produktmarke
sigma
CAS-Nummer
55976-95-9
Molekulargewicht
Test | Spezifikation |
|---|---|
| Protein by Bradford | ≥0.25 mg/ml |
| Enzyme Activity | ≥0.4 |
| µmole/mg protein Unit definition: One unit | |
| is defined as the amount of enzyme required to | |
| produce 1 umol of NADP per minute at 37C in 50mM | |
| potassium phosphate pH 7.2 containing 250uM NADPH | |
| and 25uM 9,10-phenanthrenequinone. | |
| Purity by SDS-PAGE | ≥90 % |
| Formulation | Conforms |
| Solution in 100 mM sodium phosphate, pH 7.2, | |
| containing 20% glycerol, 100 mM NaCl, and 1 mM | |
| EDTA |