Produktname
Fetales Kälberserum Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Produktnummer
f9665
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| SOURCE | Not of USA-Origin |
| APPEARANCE | Clear straw to amber liquid |
| PH | 6.7 - 8.0 |
| OSMOLALITY | 260 - 340 mOsm/Kg |
| STERILITY | pass |
| PROTEIN CONTENT | 30 - 45 g/l |
| ENDOTOXINS | ≤ 10 EU/ml |
| HAEMOGLOBIN | ≤ 25 mg/100ml |
| MYCOPLASMA | not detected |
| VIRUS (BVD) | tested |
| VIRUS (IBR, PI3) | not detected |
| CELL CULTURE TEST | pass |
| CELL LINES | report cell lines used |
| CLONING ASSAY | pass |
| ALBUMIN | report result |
| ALPHA GLOBULINS | report result |
| BETA GLOBULINS | report result |
| GAMMA GLOBULINS | report result |
| IGG | report result |
| EXPIRY PERIOD | 60 MONTHS |