Produktname
cis-4-Hydroxy-D-prolin
Produktnummer
h5877
Produktmarke
sigma
CAS-Nummer
2584-71-6
Molekulargewicht
131.13
Test | Spezifikation |
|---|---|
| Appearance (Color) | White |
| Appearance (Form) | Powder |
| Solubility (Color) | Colorless |
| Solubility (Turbidity) | Clear |
| at 100 mg/ml of H2O | |
| Infrared spectrum | Conforms to Structure |
| Carbon | 44.9 - 46.7 % |
| Nitrogen | 10.5 - 10.9 % |
| Specific Rotation | 57.0 - 59.5 ° |
| (c = 2 in H2O at 25 deg C) | |
| Purity (TLC) | ≥98 % |