Produktname
L-(+)-Milchsäure Silbersalz ~90%
Produktnummer
l7771
Produktmarke
sigma
CAS-Nummer
80298-33-5
Molekulargewicht
196.94
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | WHITE TO GRAY WITH YELLOW CAST |
| APPEARANCE (FORM) | Powder |
| SOLUBILITY (COLOR) | COLORLESS TO LIGHT YELLOW-GRAY |
| SOLUBILITY (TURBIDITY) | CLEAR TO VERY HAZY (PARTICULATES MAY BE PRESENT) |
| SOLUBILITY (METHOD) | 10 MG/ML IN WATER |
| LOSS ON DRYING | ≤ 7.0 % |
| CARBON CONTENT | 16.3 - 19.1 % * |
| ASSAY (ICP) | APPROX. 90 % (BASED ON SILVER CONTENT) |
| REMARKS | *ANHYDROUS BASIS |