Produktname
Myosin, Calcium activated from rabbit muscle buffered aqueous glycerol solution, 0.5-1.5 units/mg protein (biuret)
Produktnummer
m1636
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE | CLEAR TO HAZY COLORLESS TO LIGHT YELLOW TO LIGHT TAN LIQUID |
| PROTEIN BY BIURET | > OR = 4.0 MG/ML |
| ATPASE ACTIVITY | 0.5 TO 2.5UNITS/MG PROTEIN |
| UNIT DEFINITION | ONE UNIT WILL LIBERATE 1.0MICROMOLE OF INORGANIC PHOSPHORUS FROM ATP PER MINUTE AT PH9.0 AT 25DEGC IN THE PRESENCE OF CALCIUM. |
| DOCUMENT # M1636/12/15/09/3 |