Produktname
2-Mercaptoethanol suitable for electrophoresis
Produktnummer
m7154
Produktmarke
sigma
CAS-Nummer
60-24-2
Molekulargewicht
78.13
Test | Spezifikation |
|---|---|
| Appearance (Clarity) | Clear |
| Appearance (Color) | Colorless to Very Faint Yellow |
| Appearance (Form) | Liquid |
| Lead (Pb) | ≤1 ppm |
| Copper (Cu) | ≤1 ppm |
| Iron (Fe) | ≤1 ppm |
| Zinc (Zn) | ≤1 ppm |
| Suitability | Suitable |
| Suitable For Use in Electrophoresis | |
| Purity (GC) | ≥98.0 % |
| Recommended Retest Period | ------------------------- |
| 3 Years |