Produktname
Rabbit anti-Phospho KAP-1 (S824) Antibody, Affinity Purified Powered by Bethyl Laboratories, Inc.
Produktnummer
pla0140
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Appearance | Report Result |
| Concentration | Report Result |
| Western Blot | 1:2,000-1:10,000 |
| Immunoprecipitation | 2-10 µg/mg lysate |
| Immunohistochemistry | 1:500 to 1:2,000. Epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. |
| Immunocytochemistry | 1:200-1:1,000 |
| Immunofluorescence(ICC) | 1:200-1:1,000 |