Produktname
Spermin ≥97%
Produktnummer
s3256
Produktmarke
sigma
CAS-Nummer
71-44-3
Molekulargewicht
202.34
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Solid or Semi-solid |
| TITRATION (T) HCL 1M | 97.0 - 103.0 % |
| PURITY (GC AREA %) | ≥ 99.0 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 50MG/ML WATER |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| RECOMMENDED RETEST PERIOD | 36 MONTHS |