Produktname
(S)-(+)-Nirvanol
Produktnummer
uc180
Produktmarke
sigma
CAS-Nummer
65567-34-2
Molekulargewicht
204.23
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO OFF-WHITE CRYSTALLINE SOLID |
| H-NMR SPECTRUM | CONSISTENT WITH STRUCTURE |
| PURITY BY HPLC | MINIMUM 97% |
| SPECIFIC ROTATION | MINIMUM +108 DEG (C=0.5,ETHANOL AT 20 DEG C) |
| DOCUMENT # | UC180/10/2/06/2 |