Direkt zum Inhalt
Merck
Interleukin-4 from mouse IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Interleukin-4 from mouse

Interleukin-4 from mouse IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Synonym(e):
Interleukin-4, mIL-4, Interleukin-4 human

Products

Produktkategorie
Marke
Biologische Quelle
Physikalische Form
Reinheit
Verfahren
Showing 1-1 of 1