Direkt zum Inhalt
Merck
NT-3 human recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

NT-3 human

NT-3 human recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(e):
HGNF, NGF-2, Nerve growth factor-2, Neurotrophic factor, Neurotrophin-3

Products

Produktkategorie
Marke
Biologische Quelle
Farbe
Physikalische Form
Reinheit
Verfahren