Direkt zum Inhalt
Merck
PRMT5/MEP50 Active human recombinant, expressed in FreeStyle™ 293-F cells, ≥60% (SDS-PAGE)

PRMT5/MEP50 Active human

PRMT5/MEP50 Active human recombinant, expressed in FreeStyle™ 293-F cells, ≥60% (SDS-PAGE)
Synonym(e):
Arginine methyltransferase 5, HMT1 hnRNP methyltransferase-like 5(HRMT1L5), ICln-binding protein (IBP72) 72 kDa, Jak-binding protein 1 (JBP1), shk1 kinase-binding protein 1 homolog (SKB1)

Products

Produktkategorie
Marke
Biologische Quelle
Hersteller
Physikalische Form
Reinheit
Anwendung
Verfahren