Merck
HomeSupportCalculators & AppsBuffer Calculator
Buffer Calculator
About Biological Buffers
Buffers are used in many types of scientific studies to maintain or control the acidity of a solution within a desired physiological range, or even to initiate a particular reaction. The pH of substances affects both the rate and efficiency of chemical reactions, and the recovery and purity of products. Hence, buffers considerably contribute to variations in the outcome, and ultimately the success or failure, of studies. Therefore, understanding buffers and error-free calculation of buffers is essential to reduce variability and undesired interactions for reliable research.
Find your pH balance with our biological buffer systems.
Using the buffer preparation calculator
This buffer calculator provides an easy-to-use tool to calculate buffer molarity and prepare buffer solutions using the formula weight of the reagent and your desired volume (L, mL, or µL) and concentration (M, mM, or nM).
To calculate the amount of buffer needed, please select a buffer from the Selection menu. The empirical formula, pKa, buffer pH range, formula weight and product list will appear. Enter the desired final volume and concentration and click “Calculate Mass.” The exact mass of the buffer will be calculated in grams and a step-by-step buffer recipe is automatically provided to assist in buffer preparation.
Click on the list of product results for detailed information on our available buffers and to place an order.
CAS #:
Empirical Formula:
pKa:
Buffer pH Range:
Formula Weight (g/mol):
Formula weight
g/mol
Desired final volume
Desired concentration
  Mass=   g
Error with Formula Weight
Please check Desired Final Volume
Please check Desired Concentration
Please make a selection
Results based on your selection:
Your stock solution of is calculated to be based on the formula weight of g/mol.
To make a solution, add g of to deionized water.
Adjust the final volume of the solution to with deionized water.
Adjust the pH as needed
Buffer Calculations: Formula and Equations
  • Molar solution equation: desired molarity × formula weight × solution final volume (L) = grams needed
  • Percentage by weight (w/v): (% buffer desired / 100) × final buffer volume (mL) = g of starting material needed
  • Henderson-Hasselbach equation: pH = pKa + log [A-]/[HA]
The Henderson-Hasselbalch equation enables determination of a buffer solution's pH when the pKa is known.1 A buffer solution consists of an acid and a salt of the conjugate base of the acid. If the pH and pKa are known, the amount of salt (A-) and acid (HA) can be calculated. When the amount of salt is equal to the acid, the pH is equal to pKa and max buffering capacity is obtained. 
Reference
1.
Henderson LJ. 1908. CONCERNING THE RELATIONSHIP BETWEEN THE STRENGTH OF ACIDS AND THEIR CAPACITY TO PRESERVE NEUTRALITY. American Journal of Physiology-Legacy Content. 21(2):173-179. https://doi.org/10.1152/ajplegacy.1908.21.2.173