Merck
CDDO and Its Role in Chronic Diseases.
Advances in experimental medicine and biology (2016-10-25)
Bryan J Mathis, Taixing Cui
ZUSAMMENFASSUNG
There has been a continued interest in translational research focused on both natural products and manipulation of functional groups on these compounds to create novel derivatives with higher desired activities. Oleanolic acid, a component of traditional Chinese medicine used in hepatitis therapy, was modified by chemical processes to form 2-cyano-3,12-dioxoolean-1,9-dien-28-oic acid (CDDO). This modification increased anti-inflammatory activity significantly and additional functional groups on the CDDO backbone have shown promise in treating conditions ranging from kidney disease to obesity to diabetes. CDDO's therapeutic effect is due to its upregulation of the master antioxidant transcription factor Nuclear factor erythroid 2-related factor 2 (Nrf2) through conformational change of Nrf2-repressing, Kelch-like erythroid cell-derived protein with CNC homology-associated protein 1 (Keap1) and multiple animal and human studies have verified subsequent activation of Nrf2-controlled antioxidant genes via upstream Antioxidant Response Element (ARE) regions. At the present time, positive results have been obtained in the laboratory and clinical trials with CDDO derivatives treating conditions such as lung injury, inflammation and chronic kidney disease. However, clinical trials for cancer and cardiovascular disease have not shown equally positive results and further exploration of CDDO and its derivatives is needed to put these shortcomings into context for the purpose of future therapeutic modalities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano Supelguard Kartusche, 5 μm particle size, L × I.D. 2 cm × 4 mm