Merck
  • Development and validation of a stability-indicating RP-HPLC method for the determination of paracetamol with dantrolene or/and cetirizine and pseudoephedrine in two pharmaceutical dosage forms.
Development and validation of a stability-indicating RP-HPLC method for the determination of paracetamol with dantrolene or/and cetirizine and pseudoephedrine in two pharmaceutical dosage forms.
Talanta (2009-07-29)
Ghada M Hadad, Samy Emara, Waleed M M Mahmoud
ZUSAMMENFASSUNG
A stability-indicating reversed-phase high-performance liquid chromatography (RP-HPLC) method has been developed which can separate and accurately quantitate paracetamol, dantrolene, cetirizine and pseudoephedrine. The method was successfully validated for the purpose of conducting stability studies of the four analytes in quality control (QC) laboratories. The stability-indicating capability of the method was demonstrated by adequate separation of these four analytes from all the degradant peaks. A gradient mobile phase system consisting of (A) 50 mmol L(-1) sodium dihydrogen phosphate, 5 mmol L(-1) heptane sulfonic acid sodium salt, pH 4.2 and (B) acetonitrile was used with Discovery reversed-phase HS C(18) analytical column (250 mm x 4.6 mm i.d., 5 microm particle size). Quantitation was achieved with UV detection at 214 nm, based on peak area. The proposed method was validated and successfully applied for the analysis of pharmaceutical formulations and laboratory-prepared mixtures containing the two multicomponent combinations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Supelco
Discovery® C18 Supelguard Vorsäulen-Kartusche, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Supelco
Discovery® C18 Supelguard Vorsäulen-Kartusche, Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 4 mm
Supelco
Discovery® C18 Supelguard Vorsäulen-Kartusche, Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C18 Supelguard Vorsäulen-Kartusche, 5 μm particle size, L × I.D. 2 cm × 3 mm, pkg of 2 ea
Supelco
Discovery® C18 Supelguard Vorsäulen-Kartusche, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea
Supelco
Discovery® C18 Supelguard Vorsäulen-Kartusche, Kit, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
Discovery® C18 HPLC-Säule, 5 μm particle size, L × I.D. 12.5 cm × 2.1 mm