Merck
  • Mitogen-activated protein kinase 14 is a novel negative regulatory switch for the vascular smooth muscle cell contractile gene program.
Mitogen-activated protein kinase 14 is a novel negative regulatory switch for the vascular smooth muscle cell contractile gene program.
Arteriosclerosis, thrombosis, and vascular biology (2012-11-24)
Xiaochun Long, Sarah L Cowan, Joseph M Miano
ZUSAMMENFASSUNG
Several studies have shown through chemical inhibitors that p38 mitogen-activated protein kinase (MAPK) promotes vascular smooth muscle cell (VSMC) differentiation. Here, we evaluate the effects of knocking down a dominant p38MAPK isoform on VSMC differentiation. Knockdown of p38MAPKα (MAPK14) in human coronary artery SMCs unexpectedly increases VSMC differentiation genes, such as miR145, ACTA2, CNN1, LMOD1, and TAGLN, with little change in the expression of serum response factor (SRF) and 2 SRF cofactors, myocardin (MYOCD) and myocardin-related transcription factor A (MKL1). A variety of chemical and biological inhibitors demonstrate a critical role for a RhoA-MKL1-SRF-dependent pathway in mediating these effects. MAPK14 knockdown promotes MKL1 nuclear localization and VSMC marker expression, an effect partially reversed with Y27632; in contrast, MAP2K6 (MKK6) blocks MKL1 nuclear import and VSMC marker expression. Immunostaining and Western blotting of injured mouse carotid arteries reveal elevated MAPK14 (both total and phosphorylated) and reduced VSMC marker expression. Reduced MAPK14 expression evokes unanticipated increases in VSMC contractile genes, suggesting an unrecognized negative regulatory role for MAPK14 signaling in VSMC differentiation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid