Merck
Fast label-free cytoskeletal network imaging in living mammalian cells.
Biophysical journal (2014-04-18)
Pierre Bon, Sandrine Lécart, Emmanuel Fort, Sandrine Lévêque-Fort
ZUSAMMENFASSUNG
We present a full-field technique that allows label-free cytoskeletal network imaging inside living cells. This noninvasive technique allows monitoring of the cytoskeleton dynamics as well as interactions between the latter and organelles on any timescale. It is based on high-resolution quantitative phase imaging (modified Quadriwave lateral shearing interferometry) and can be directly implemented using any optical microscope without modification. We demonstrate the capability of our setup on fixed and living Chinese hamster ovary cells, showing the cytoskeleton dynamics in lamellipodia during protrusion and mitochondria displacement along the cytoskeletal network. In addition, using the quantitative function of the technique, along with simulation tools, we determined the refractive index of a single tubulin microtubule to be ntubu=2.36±0.6 at λ=527 nm.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture