Merck
  • Antinociceptive action of NOP and opioid receptor agonists in the mouse orofacial formalin test.
Antinociceptive action of NOP and opioid receptor agonists in the mouse orofacial formalin test.
Peptides (2017-07-13)
A Rizzi, C Ruzza, S Bianco, C Trapella, G Calo'
ZUSAMMENFASSUNG
Nociceptin/orphanin FQ (N/OFQ) modulates several biological functions, including pain transmission via selective activation of a specific receptor named NOP. The aim of this study was the investigation of the antinociceptive properties of NOP agonists and their interaction with opioids in the trigeminal territory. The orofacial formalin (OFF) test in mice was used to investigate the antinociceptive potential associated to the activation of NOP and opioid receptors. Mice subjected to OFF test displayed the typical biphasic nociceptive response and sensitivity to opioid and NSAID drugs. Mice knockout for the NOP gene displayed a robust pronociceptive phenotype. The NOP selective agonist Ro 65-6570 (0.1-1mgkg
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ro 65-6570 Hydrochloride, ≥98% (HPLC)