Merck
  • Pharmacological characterization of cebranopadol a novel analgesic acting as mixed nociceptin/orphanin FQ and opioid receptor agonist.
Pharmacological characterization of cebranopadol a novel analgesic acting as mixed nociceptin/orphanin FQ and opioid receptor agonist.
Pharmacology research & perspectives (2017-01-25)
Anna Rizzi, Maria Camilla Cerlesi, Chiara Ruzza, Davide Malfacini, Federica Ferrari, Sara Bianco, Tommaso Costa, Remo Guerrini, Claudio Trapella, Girolamo Calo'
ZUSAMMENFASSUNG
The aim of the study was to investigate the in vitro and in vivo pharmacological profile of cebranopadol, a novel agonist for opioid and nociceptin/orphanin FQ (N/OFQ) receptors (NOP). In vitro cebranopadol was assayed in calcium mobilization studies in cells coexpressing NOP or opioid receptors and chimeric G-proteins and in a bioluminescence resonance energy transfer (BRET) assay for studying receptor interaction with G-protein and
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ro 65-6570 Hydrochloride, ≥98% (HPLC)