Merck
  • Glucose Starvation Increases V-ATPase Assembly and Activity in Mammalian Cells through AMP
    Kinase and Phosphatidylinositide 3-Kinase/Akt Signaling
Glucose Starvation Increases V-ATPase Assembly and Activity in Mammalian Cells through AMP<BR/> Kinase and Phosphatidylinositide 3-Kinase/Akt Signaling
The Journal of Biological Chemistry (2018)
Christina M. McGuire and Michael Forgac