Merck
  • Asparagine attenuates intestinal injury, improves energy status and inhibits AMP-activated protein kinase signalling pathways in weaned piglets challenged with Escherichia coli lipopolysaccharide.
Asparagine attenuates intestinal injury, improves energy status and inhibits AMP-activated protein kinase signalling pathways in weaned piglets challenged with Escherichia coli lipopolysaccharide.
The British journal of nutrition (2015-08-19)
Xiuying Wang, Yulan Liu, Shuang Li, Dingan Pi, Huiling Zhu, Yongqing Hou, Haifeng Shi, Weibo Leng
ZUSAMMENFASSUNG
The intestine requires a high amount of energy to maintain its health and function; thus, energy deficits in intestinal mucosa may lead to intestinal damage. Asparagine (Asn) is a precursor for many other amino acids such as aspartate, glutamine and glutamate, which can be used to supply energy to enterocytes. In the present study, we hypothesise that dietary supplementation of Asn could alleviate bacterial lipopolysaccharide (LPS)-induced intestinal injury via improvement of intestinal energy status. A total of twenty-four weaned piglets were assigned to one of four treatments: (1) non-challenged control; (2) LPS+0 % Asn; (3) LPS+0·5 % Asn; (4) LPS+1·0 % Asn. On day 19, piglets were injected with LPS or saline. At 24 h post-injection, piglets were slaughtered and intestinal samples were collected. Asn supplementation improved intestinal morphology, indicated by higher villus height and villus height:crypt depth ratio, and lower crypt depth. Asn supplementation also increased the ratios of RNA:DNA and protein:DNA as well as disaccharidase activities in intestinal mucosa. In addition, Asn supplementation attenuated bacterial LPS-induced intestinal energy deficits, indicated by increased ATP and adenylate energy charge levels, and decreased AMP:ATP ratio. Moreover, Asn administration increased the activities of key enzymes involved in the tricarboxylic acid cycle, including citrate synthase, isocitrate dehydrogenase and α-ketoglutarate dehydrogenase complex. Finally, Asn administration decreased the mRNA abundance of intestinal AMP-activated protein kinase-α1 (AMPKα1), AMPKα2, silent information regulator 1 (SIRT1) and PPARγ coactivator-1α (PGC1α), and reduced intestinal AMPKα phosphorylation. Collectively, these results indicate that Asn supplementation alleviates bacterial LPS-induced intestinal injury by modulating the AMPK signalling pathway and improving energy status.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution