Merck
  • Opposite Effects of SET7/9 on Apoptosis of Human Acute Myeloid Leukemia Cells and Lung Cancer Cells.
Opposite Effects of SET7/9 on Apoptosis of Human Acute Myeloid Leukemia Cells and Lung Cancer Cells.
Journal of Cancer (2017-08-19)
Ye Gu, Yuan Wang, Xinling Wang, Lili Gao, Weiping Yu, Wei-Feng Dong
ZUSAMMENFASSUNG
SET7/9 is a protein lysine methyltransferases (PLMTs or PKMTs) which methylates both histone H3K4 and non-histone proteins including transcriptional factors, tumor suppressors, and membrane-associated receptors. Methylation of these proteins alters protein activity and leads to changes in cellular behavior and a series of biological processes. This study aims to investigate the role of SET7/9 in human acute myeloid leukemia (AML) and non-small-cell lung cancer (NSCLC). We examined the expression of SET7/9 in AML cells and NSCLC cells and detected the methylation status of the SET7/9 promoter region. To evaluate the effect of SET7/9 expression changes on cell apoptosis, cell apoptosis rates were determined after SET7/9 overexpression or down-regulation. Our results showed that SET7/9 induces apoptosis of AML cells and inhibits apoptosis of NSCLC cells, suggesting differential effects of SET7/9 on cellular apoptosis and carcinogenesis depending on different cancer types and genetic contexts. Furthermore, we also demonstrated that SET7/9 suppresses cell apoptosis via modulation of E2F1 under circumstance of p53 deficiency in NSCLC cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid