Merck
  • Reciprocal synapses between mushroom body and dopamine neurons form a positive feedback loop required for learning.
Reciprocal synapses between mushroom body and dopamine neurons form a positive feedback loop required for learning.
eLife (2017-05-11)
Isaac Cervantes-Sandoval, Anna Phan, Molee Chakraborty, Ronald L Davis
ZUSAMMENFASSUNG
Current thought envisions dopamine neurons conveying the reinforcing effect of the unconditioned stimulus during associative learning to the axons of Drosophila mushroom body Kenyon cells for normal olfactory learning. Here, we show using functional GFP reconstitution experiments that Kenyon cells and dopamine neurons from axoaxonic reciprocal synapses. The dopamine neurons receive cholinergic input via nicotinic acetylcholine receptors from the Kenyon cells; knocking down these receptors impairs olfactory learning revealing the importance of these receptors at the synapse. Blocking the synaptic output of Kenyon cells during olfactory conditioning reduces presynaptic calcium transients in dopamine neurons, a finding consistent with reciprocal communication. Moreover, silencing Kenyon cells decreases the normal chronic activity of the dopamine neurons. Our results reveal a new and critical role for positive feedback onto dopamine neurons through reciprocal connections with Kenyon cells for normal olfactory learning.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-grünfluoreszierendes Protein (GFP)-Antikörper in Maus hergestellte Antikörper, clone GFP-20, ascites fluid