Merck
SNX18 regulates ATG9A trafficking from recycling endosomes by recruiting Dynamin-2.
EMBO reports (2018-02-14)
Kristiane Søreng, Michael J Munson, Christopher A Lamb, Gunnveig T Bjørndal, Serhiy Pankiv, Sven R Carlsson, Sharon A Tooze, Anne Simonsen
ZUSAMMENFASSUNG
Trafficking of mammalian ATG9A between the Golgi apparatus, endosomes and peripheral ATG9A compartments is important for autophagosome biogenesis. Here, we show that the membrane remodelling protein SNX18, previously identified as a positive regulator of autophagy, regulates ATG9A trafficking from recycling endosomes. ATG9A is recruited to SNX18-induced tubules generated from recycling endosomes and accumulates in juxtanuclear recycling endosomes in cells lacking SNX18. Binding of SNX18 to Dynamin-2 is important for ATG9A trafficking from recycling endosomes and for formation of ATG16L1- and WIPI2-positive autophagosome precursor membranes. We propose a model where upon autophagy induction, SNX18 recruits Dynamin-2 to induce budding of ATG9A and ATG16L1 containing membranes from recycling endosomes that traffic to sites of autophagosome formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor
Sigma-Aldrich
3-Hydroxy-1,2-dimethyl-4(1H)-pyridon, 98%