Merck
Substrate Stiffness Combined with Hepatocyte Growth Factor Modulates Endothelial Cell Behavior.
Biomacromolecules (2016-07-19)
Hao Chang, Xi-Qiu Liu, Mi Hu, He Zhang, Bo-Chao Li, Ke-Feng Ren, Thomas Boudou, Corinne Albiges-Rizo, Catherine Picart, Jian Ji
ZUSAMMENFASSUNG
Endothelial cells (ECs) play a crucial role in regulating various physiological and pathological processes. The behavior of ECs is modulated by physical (e.g., substrate stiffness) and biochemical cues (e.g., growth factors). However, the synergistic influence of these cues on EC behavior has rarely been investigated. In this study, we constructed poly(l-lysine)/hyaluronan (PLL/HA) multilayer films with different stiffness and exposed ECs to these substrates with and without hepatocyte growth factor (HGF)-supplemented culture medium. We demonstrated that EC adhesion, migration, and proliferation were positively correlated with substrate stiffness and that these behaviors were further promoted by HGF. Interestingly, ECs on the lower stiffness substrates showed stronger responses to HGF in terms of migration and proliferation, suggesting that HGF can profoundly influence stiffness-dependent EC behavior correlated with EC growth. After the formation of an EC monolayer, EC behaviors correlated with endothelial function were evaluated by characterizing monolayer integrity, nitric oxide production, and gene expression of endothelial nitric oxide synthase. For the first time, we demonstrated that endothelial function displayed a negative correlation with substrate stiffness. Although HGF improved endothelial function, HGF was not able to change the stiffness-dependent manner of endothelial functions. Taken together, this study provides insights into the synergetic influence of physical and biochemical cues on EC behavior and offers great potential in the development of optimized biomaterials for EC-based regenerative medicine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-CD31(PECAM-1) antibody , Mouse monoclonal, clone WM-59, purified from hybridoma cell culture