Merck
  • Chroman-4-one- and chromone-based sirtuin 2 inhibitors with antiproliferative properties in cancer cells.
Chroman-4-one- and chromone-based sirtuin 2 inhibitors with antiproliferative properties in cancer cells.
Journal of medicinal chemistry (2014-11-11)
Tina Seifert, Marcus Malo, Tarja Kokkola, Karin Engen, Maria Fridén-Saxin, Erik A A Wallén, Maija Lahtela-Kakkonen, Elina M Jarho, Kristina Luthman
ZUSAMMENFASSUNG
Sirtuins (SIRTs) catalyze the NAD(+)-dependent deacetylation of N(ε)-acetyl lysines on various protein substrates. SIRTs are interesting drug targets as they are considered to be related to important pathologies such as inflammation and aging-associated diseases. We have previously shown that chroman-4-ones act as potent and selective inhibitors of SIRT2. Herein we report novel chroman-4-one and chromone-based SIRT2 inhibitors containing various heterofunctionalities to improve pharmacokinetic properties. The compounds retained both high SIRT2 selectivity and potent inhibitory activity. Two compounds were tested for their antiproliferative effects in breast cancer (MCF-7) and lung carcinoma (A549) cell lines. Both compounds showed antiproliferative effects correlating with their SIRT2 inhibition potency. They also increased the acetylation level of α-tubulin, indicating that SIRT2 is likely to be the target in cancer cells. A binding mode of the inhibitors that is consistent with the SAR data was proposed based on a homology model of SIRT2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monoklonales Anti-Tubulin, acetyliert in Maus hergestellte Antikörper, clone 6-11B-1, ascites fluid