Merck
UBE3B Is a Calmodulin-regulated, Mitochondrion-associated E3 Ubiquitin Ligase.
The Journal of biological chemistry (2016-12-23)
Andrea Braganza, Jianfeng Li, Xuemei Zeng, Nathan A Yates, Nupur B Dey, Joel Andrews, Jennifer Clark, Leila Zamani, Xiao-Hong Wang, Claudette St Croix, Roderick O'Sullivan, Laura Garcia-Exposito, Jeffrey L Brodsky, Robert W Sobol
ZUSAMMENFASSUNG
Recent genome-wide studies found that patients with hypotonia, developmental delay, intellectual disability, congenital anomalies, characteristic facial dysmorphic features, and low cholesterol levels suffer from Kaufman oculocerebrofacial syndrome (KOS, also reported as blepharophimosis-ptosis-intellectual disability syndrome). The primary cause of KOS is autosomal recessive mutations in the gene UBE3B However, to date, there are no studies that have determined the cellular or enzymatic function of UBE3B. Here, we report that UBE3B is a mitochondrion-associated protein with homologous to the E6-AP Cterminus (HECT) E3 ubiquitin ligase activity. Mutating the catalytic cysteine (C1036A) or deleting the entire HECT domain (amino acids 758-1068) results in loss of UBE3B's ubiquitylation activity. Knockdown of UBE3B in human cells induces changes in mitochondrial morphology and physiology, a decrease in mitochondrial volume, and a severe suppression of cellular proliferation. We also discovered that UBE3B interacts with calmodulin via its N-terminal isoleucine-glutamine (IQ) motif. Deletion of the IQ motif (amino acids 29-58) results in loss of calmodulin binding and a significant increase in the in vitro ubiquitylation activity of UBE3B. In addition, we found that changes in calcium levels in vitro disrupt the calmodulin-UBE3B interaction. These studies demonstrate that UBE3B is an E3 ubiquitin ligase and reveal that the enzyme is regulated by calmodulin. Furthermore, the modulation of UBE3B via calmodulin and calcium implicates a role for calcium signaling in mitochondrial protein ubiquitylation, protein turnover, and disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isolierungskit für das endoplasmatische Retikulum, isolation of intact ER from mammalian soft tissues and cultured cells
Roche
Anti-HA-Affinitätsmatrix, from rat IgG1
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-UBE3B antibody produced in rabbit, affinity isolated antibody