Merck
  • Histone deacetylase inhibition enhances antimicrobial peptide but not inflammatory cytokine expression upon bacterial challenge.
Histone deacetylase inhibition enhances antimicrobial peptide but not inflammatory cytokine expression upon bacterial challenge.
Proceedings of the National Academy of Sciences of the United States of America (2016-05-11)
Natalie Fischer, Emmanuel Sechet, Robin Friedman, Aurélien Amiot, Iradj Sobhani, Giulia Nigro, Philippe J Sansonetti, Brice Sperandio
ZUSAMMENFASSUNG
Antimicrobial peptides (AMP) are defense effectors of the innate immunity playing a crucial role in the intestinal homeostasis with commensals and protection against pathogens. Herein we aimed to investigate AMP gene regulation by deciphering specific characteristics allowing their enhanced expression among innate immune genes, particularly those encoding proinflammatory mediators. Our emphasis was on epigenetic regulation of the gene encoding the AMP β-defensin 2 (HBD2), taken as a model of possibly specific induction, upon challenge with a commensal bacterium, compared with the proinflammatory cytokine IL-8. Using an in vitro model of colonic epithelial cells challenged with Escherichia coli K12, we showed that inhibition of histone deacetylases (HDAC) by trichostatin A dramatically enhanced induction of HBD2 expression, without affecting expression of IL-8. This mechanism was supported by an increased phosphorylation of histone H3 on serine S10, preferentially at the HBD2 promoter. This process occurred through activation of the IκB kinase complex, which also led to activation of NF-κB. Moreover, we demonstrated that NF-κB was modified by acetylation upon HDAC inhibition, partly by the histone acetyltransferase p300, and that both NF-κB and p300 supported enhanced induction of HBD2 expression. Furthermore, we identified additional genes belonging to antimicrobial defense and epithelial restitution pathways that showed a similar pattern of epigenetic control. Finally, we confirmed our finding in human colonic primary cells using an ex vivo organoid model. This work opens the way to use epigenetic pharmacology to achieve induction of epithelial antimicrobial defenses, while limiting the deleterious risk of an inflammatory response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.
Sigma-Aldrich
Anti-Acetyl-Histon-H3(Lys14)-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Acetyl-Histon-H4-Antikörper, serum, Upstate®
Sigma-Aldrich
5-Azacytidin, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Anti-Acetyl-Histone-H3(Lys23)-Antikörper, serum, Upstate®
Sigma-Aldrich
C646, ≥98% (HPLC)
Sigma-Aldrich
BMS-345541, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Acetyl-Histon-H4(Lys5)-Antikörper, serum, from rabbit
Sigma-Aldrich
Anti-Acetyl-Histon-H4(Lys12)-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Acetyl-Histon-H4(Lys8)-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Acetyl-Histon-H3(Lys56)-Antikörper, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Acetyl-Histon-H3(Lys 4)-Antikörper, Upstate®, from rabbit