Merck
  • A-Kinase Anchor Protein 12 Is Required for Oligodendrocyte Differentiation in Adult White Matter.
A-Kinase Anchor Protein 12 Is Required for Oligodendrocyte Differentiation in Adult White Matter.
Stem cells (Dayton, Ohio) (2018-01-10)
Takakuni Maki, Yoon Kyung Choi, Nobukazu Miyamoto, Akihiro Shindo, Anna C Liang, Bum Ju Ahn, Emiri T Mandeville, Seiji Kaji, Kanako Itoh, Ji Hae Seo, Irwin H Gelman, Josephine Lok, Ryosuke Takahashi, Kyu-Won Kim, Eng H Lo, Ken Arai
ZUSAMMENFASSUNG
Oligodendrocyte precursor cells (OPCs) give rise to oligodendrocytes in cerebral white matter. However, the underlying mechanisms that regulate this process remain to be fully defined, especially in adult brains. Recently, it has been suggested that signaling via A-kinase anchor protein 12 (AKAP12), a scaffolding protein that associates with intracellular molecules such as protein kinase A, may be involved in Schwann cell homeostasis and peripheral myelination. Here, we asked whether AKAP12 also regulates the mechanisms of myelination in the CNS. AKAP12 knockout mice were compared against wild-type (WT) mice in a series of neurochemical and behavioral assays. Compared with WTs, 2-months old AKAP12 knockout mice exhibited loss of myelin in white matter of the corpus callosum, along with perturbations in working memory as measured by a standard Y-maze test. Unexpectedly, very few OPCs expressed AKAP12 in the corpus callosum region. Instead, pericytes appeared to be one of the major AKAP12-expressing cells. In a cell culture model system, conditioned culture media from normal pericytes promoted in-vitro OPC maturation. However, conditioned media from AKAP12-deficient pericytes did not support the OPC function. These findings suggest that AKAP12 signaling in pericytes may be required for OPC-to-oligodendrocyte renewal to maintain the white matter homeostasis in adult brain. Stem Cells 2018;36:751-760.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human AKAP12