Merck
  • Reaction kinetic studies for comparison of mutagenic potency between butadiene monoxide and glycidamide.
Reaction kinetic studies for comparison of mutagenic potency between butadiene monoxide and glycidamide.
Chemico-biological interactions (2018-04-14)
Hitesh V Motwani, Lars Eriksson, Lisa Göpfert, Kristian Larsen
ZUSAMMENFASSUNG
DNA adducts can be formed from covalent binding of electrophilic reactive compounds to the nucleophilic N- and O-atoms of the biomolecule. The O-sites on DNA, with nucleophilic strength (n) of ca. 2, is recognized as a critical site for mutagenicity. Characterization of the reactivity of electrophilic compounds at the O-sites can be used to predict their mutagenic potency in relative terms. In the present study, reaction kinetic experiments were performed for butadiene monoxide (BM) in accordance with the Swain-Scott relation using model nucleophiles representing N- and O-sites on DNA, and earlier for glycidamide (GA) using a similar approach. The epoxide from the kinetic experiments was trapped by cob(I)alamin, resulting in formation of an alkylcobalamin which was analyzed by liquid chromatography tandem mass spectrometry. The Swain-Scott relationship was used to determine selectivity constant (s) of BM and GA as 0.86 and 1.0, respectively. The rate constant for the reaction at n of 2 was extrapolated to 0.023 and 0.038 M-1 h-1 for BM and GA, respectively, implying a higher mutagenic potency per dose unit of GA compared to BM. The reaction kinetic parameters associated with mutagenic potency were also estimated by a density functional theory approach, which were in accordance to the experimental determined values. These types of reaction kinetic measures could be useful in development of a chemical reactivity based prediction tool that could aid in reduction of animal experiments in cancer risk assessment procedures for relative mutagenicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Epoxy-1-buten, 98%