Merck
Occurrence of pesticide residues in fish from south American rainfed agroecosystems.
The Science of the total environment (2018-03-11)
Federico Ernst, Beatriz Alonso, Marcos Colazzo, Lucia Pareja, Verónica Cesio, Alfredo Pereira, Alejandro Márquez, Eugenia Errico, Angel Manuel Segura, Horacio Heinzen, Andrés Pérez-Parada
ZUSAMMENFASSUNG
Environmental sustainability of South American rainfed agroecosystems is of current concern. In this work, we evaluate the occurrence of multiple pesticide residues in muscle tissue of wild fish species from two large rivers in South America (Uruguay and Negro Rivers). Two sampling campaigns (representing summer and winter crops) were performed during 2015 targeting a wide biodiversity of fish species used for human consumption (ranging from migratory to non-migratory and from detritivorous to top-predators). Three different localities associated to rainfed agriculture were assessed, two of them enclosed to a RAMSAR site (National Park "Esteros de Farrapos e Islas del Rio Uruguay"). Pesticide residues occurred in muscle tissue of 143 from 149 sampled fishes (96%). Thirty different pesticides were detected at concentrations from <1 to 194μgkg-1. Incidence of pesticides in fish were tightly related to: i) features of the contaminant: (Kow, environmental persistence and mobility) and ii) intensity of use of particular pesticides and land dedicated to rainfed agriculture. Trifloxystrobin, metolachlor and pyraclostrobin showed the highest rates of occurrence. Of great concern is that strobirulins have highest toxicity to fish from those detected compounds. From the pattern of pesticides occurring for non-migratory fish species it was possible to trend important spatial differences related to the intensity of rainfed agriculture. Results suggest a regular exposition of aquatic wild biota to sublethal concentrations of multiple semi-polar pesticides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Supelco
Bromphosmethyl, PESTANAL®, analytical standard