Merck
Synchronous immune alterations mirror clinical response during allergen immunotherapy.
The Journal of allergy and clinical immunology (2017-11-13)
Amedee Renand, Mohamed H Shamji, Kristina M Harris, Tielin Qin, Erik Wambre, Guy W Scadding, Peter A Wurtzen, Stephen J Till, Alkis Togias, Gerald T Nepom, William W Kwok, Stephen R Durham
ZUSAMMENFASSUNG
Three years of treatment with either sublingual or subcutaneous allergen immunotherapy has been shown to be effective and to induce long-term tolerance. The Gauging Response in Allergic Rhinitis to Sublingual and Subcutaneous Immunotherapy (GRASS) trial demonstrated that 2 years of treatment through either route was effective in suppressing the response to nasal allergen challenge, although it was insufficient for inhibition 1 year after discontinuation. We sought to examine in the GRASS trial the time course of immunologic changes during 2 years of sublingual and subcutaneous immunotherapy and for 1 year after treatment discontinuation. We performed multimodal immunomonitoring to assess allergen-specific CD4 T-cell properties in parallel with analysis of local mucosal cytokine responses induced by nasal allergen exposure and humoral immune responses that included IgE-dependent basophil activation and measurement of serum inhibitory activity for allergen-IgE binding to B cells (IgE-facilitated allergen binding). All 3 of these distinct arms of the immune response displayed significant and coordinate alterations during 2 years of allergen desensitization, followed by reversal at 3 years, reflecting a lack of a durable immunologic effect. Although frequencies of antigen-specific TH2 cells in peripheral blood determined by using HLA class II tetramer analysis most closely paralleled clinical outcomes, IgE antibody-dependent functional assays remained inhibited in part 1 year after discontinuation. Two years of allergen immunotherapy were effective but insufficient for long-term tolerance. Allergen-specific TH2 cells most closely paralleled the transient clinical outcome, and it is likely that recurrence of the T-cell drivers of allergic immunity abrogated the potential for durable tolerance. On the other hand, the persistence of IgE blocking antibody 1 year after discontinuation might be an early indicator of a protolerogenic mechanism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dasatinib