Merck
  • Inhibition of Pathological Phenotype of Hypertrophic Scar Fibroblasts Via Coculture with Adipose-Derived Stem Cells.
Inhibition of Pathological Phenotype of Hypertrophic Scar Fibroblasts Via Coculture with Adipose-Derived Stem Cells.
Tissue engineering. Part A (2017-06-01)
Jingcheng Deng, Yuan Shi, Zhen Gao, Wenjie Zhang, Xiaoli Wu, Weigang Cao, Wei Liu
ZUSAMMENFASSUNG
Hypertrophic scar (HS) is a dermal fibroproliferative disease characterized by fibroblast over-proliferation, overproduction, and deposition of the extracellular matrix. Growing evidence demonstrated that adipose-derived stem cells (ASCs) secrete a plethora of trophic and antifibrotic factors, which suppress inflammation and ameliorate fibrosis of different tissues. However, few studies investigate their effect on repressing HS activity. This study evaluated the suppressing effect of ASCs on HS fibroblast bioactivity and the possible mechanism via a coculture model. HS-derived fibroblasts (HSFs) and ASCs were isolated from individual patients. HSFs or HSFs treated with transforming growth factor-β1 (TGF-β1) were cocultured with ASCs and the change of HSF cellular behaviors, such as cell proliferation, migration, contractility, and gene/protein expression of scar-related molecules, were evaluated by cell counting assay, cell cycle analysis, scratch wound assay, fibroblast-populated collagen lattice (FPCL) contractility assay, real-time quantitative polymerase chain reaction, ELISA, and western blotting assay. After 5 days of ASC coculture treatment, the expression levels of collagen I (Col 1), collagen III (Col 3), fibronectin (FN), TGF-β1, interleukin-6 (IL-6), interleukin-8 (IL-8), connective tissue growth factor (CTGF), and alpha-smooth muscle actin (α-SMA) in HSFs decreased significantly while the expression levels of decorin (DCN) and MMP-1/TIMP-1 (matrix metalloproteinase/tissue inhibitor of MMP) ratio increased significantly. Besides, after 5 days of exogenous TGF-β1 stimulation, the expression levels of Col 1, FN, TGF-β1, IL-6, CTGF, and α-SMA in HSFs increased significantly. Impressively, all these increased gene expression levels were reversed by 5 days of ASCs coculture treatment. Additionally, the proliferation, migration, and contractility of HSFs were all significantly reduced by ASC coculture treatment. Furthermore, the protein levels of TGF-β1 and intracellular signal pathway-related molecules, such as p-smad2, p-smad3, p-Stat3, and p-ERK, were downregulated significantly in HSFs after 5 days of ASCs coculture treatment. This study demonstrated that coculture of HSFs with ASCs not only inhibited proliferation, migration, and contractility of HSFs but also decreased the expression levels of HSF-related or TGF-β1-induced molecules. Additionally, the antifibrotic effect on HSFs was likely mediated by the inhibition of multiple intracellular signaling. The results of this study suggest the therapeutic potential of ASCs for HS treatment, which is worth of further investigation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade