Merck
  • BRD7 inhibits the Warburg effect and tumor progression through inactivation of HIF1α/LDHA axis in breast cancer.
BRD7 inhibits the Warburg effect and tumor progression through inactivation of HIF1α/LDHA axis in breast cancer.
Cell death & disease (2018-05-05)
Weihong Niu, Yanwei Luo, Xinye Wang, Yao Zhou, Hui Li, Heran Wang, Yaojie Fu, Shanshan Liu, Shanghelin Yin, Jianglei Li, Ran Zhao, Yukun Liu, Songqing Fan, Zheng Li, Wei Xiong, Xiaoling Li, Guiyuan Li, Caiping Ren, Ming Tan, Ming Zhou
ZUSAMMENFASSUNG
The bromodomain-containing protein 7 (BRD7) was first identified as a tumor suppressor in nasopharyngeal carcinoma and has critical roles in cancer development and progression. However, the regulatory roles and mechanisms of BRD7 in cancer metabolism are still unknown. In this study, we demonstrated that BRD7 was lowly expressed in breast cancer tissues and was identified as a poor prognostic factor in breast cancer. Meanwhile, BRD7 could suppress cell proliferation, initiate cell apoptosis and reduce aerobic glycolysis, suggesting that BRD7 plays a tumor suppressive roles in breast cancer. Mechanistically, BRD7 could negatively regulate a critical glycolytic enzyme LDHA through directly interaction with its upstream transcription factor, HIF1α, facilitating degradation of HIF1α mediated by ubiquitin-proteasome pathway. Moreover, restoring the expression of LDHA in breast cancer cells could reverse the effect of BRD7 on aerobic glycolysis, cell proliferation, and tumor formation, as well as the expression of cell cycle and apopotosis related molecules such as cyclin D1, CDK4, P21, and c-PARP both in vitro and in vivo. Taken together, these results indicate that BRD7 acts as a tumor suppressor in breast cancer and represses the glycolysis and tumor progression through inactivation of HIF1α/LDHA transcription axis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natrium-Oxamat, ≥98%