Merck
  • Three new species of Thelepus Leuckart, 1849 from Europe and a re-description of T. cincinnatus (Fabricius, 1780) (Annelida, Terebellidae).
Three new species of Thelepus Leuckart, 1849 from Europe and a re-description of T. cincinnatus (Fabricius, 1780) (Annelida, Terebellidae).
ZooKeys (2018-07-27)
Igor Jirkov
ZUSAMMENFASSUNG
The review of a large amount of material previously identified as the terebellid annelid, Thelepus cincinnatus (Fabricius, 1780) shows that, within European waters from the Mediterranean to the North Pole, this species should be split into four species, three of which (T. davehallisp. n., T. marthaesp. n., and T. paraparisp. n.) are newly described here and T. cincinnatus s. str. is re-described. These four species each show distinct distribution ranges. Thelepus cincinnatus has notopodia on almost all segments and numerous eyespots; it inhabits the high boreal and arctic shelf and the North Atlantic slope, and probably also occurs on the North Pacific shelf and slope. Thelepus marthaesp. n. has no eyespots and inhabits deep waters of the high Arctic. Thelepus davehallisp. n. has no eyespots and has notopodia on 1/2 to 2/3 of the anterior of the body; it inhabits boreal shelf waters (from Iceland to the Mediterranean) below the tidal front. Thelepus paraparisp. n. differs from the previous three species in that the uncini of the first uncinigerous segment has two teeth above the main fang; it inhabits shallow, coastal waters of the Mediterranean, inshore from the tidal front.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)