Merck
  • LysMD3 is a type II membrane protein without an in vivo role in the response to a range of pathogens.
LysMD3 is a type II membrane protein without an in vivo role in the response to a range of pathogens.
The Journal of biological chemistry (2018-03-03)
Christine C Yokoyama, Megan T Baldridge, Daisy W Leung, Guoyan Zhao, Chandni Desai, Ta-Chiang Liu, Vladimir E Diaz-Ochoa, Jeremy P Huynh, Jacqueline M Kimmey, Erica L Sennott, Camaron R Hole, Rachel A Idol, Sunmin Park, Kelly M Storek, Caihong Wang, Seungmin Hwang, Ashley Viehmann Milam, Eric Chen, Tobias Kerrinnes, Michael N Starnbach, Scott A Handley, Indira U Mysorekar, Paul M Allen, Denise M Monack, Mary C Dinauer, Tamara L Doering, Renee M Tsolis, Jonathan E Dworkin, Christina L Stallings, Gaya K Amarasinghe, Craig A Micchelli, Herbert W Virgin
ZUSAMMENFASSUNG
Germline-encoded receptors recognizing common pathogen-associated molecular patterns are a central element of the innate immune system and play an important role in shaping the host response to infection. Many of the innate immune molecules central to these signaling pathways are evolutionarily conserved. LysMD3 is a novel molecule containing a putative peptidoglycan-binding domain that has orthologs in humans, mice, zebrafish, flies, and worms. We found that the lysin motif (LysM) of LysMD3 is likely related to a previously described peptidoglycan-binding LysM found in bacteria. Mouse LysMD3 is a type II integral membrane protein that co-localizes with GM130+ structures, consistent with localization to the Golgi apparatus. We describe here two lines of mLysMD3-deficient mice for in vivo characterization of mLysMD3 function. We found that mLysMD3-deficient mice were born at Mendelian ratios and had no obvious pathological abnormalities. They also exhibited no obvious immune response deficiencies in a number of models of infection and inflammation. mLysMD3-deficient mice exhibited no signs of intestinal dysbiosis by 16S analysis or alterations in intestinal gene expression by RNA sequencing. We conclude that mLysMD3 contains a LysM with cytoplasmic orientation, but we were unable to define a physiological role for the molecule in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Anti-Histon H2A.X-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-LYSMD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution